Ethyl 3-amino-2,2-dimethylpropanoate hydrochloride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:80253-38-9
:
Formula:C₇H₁₅NO₂·ClH
MDL Number:MFCD00102231
Molecular Weight:181.66 g/mol
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together