7-Azaspiro[3.5]nonan-2-ol hydrochloride
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: 7-azaspiro[3.5]nonan-2-ol hydrochloride
CAS Number:587869-08-7
:
Formula:C₈H₁₅NO·ClH
MDL Number:MFCD20486137
Molecular Weight:177.672 g/mol
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together