Methyl 2-chloro-5-cyanobenzoate 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:914106-36-8
:
Formula:C₉H₆ClNO₂
MDL Number:MFCD13192027
Molecular Weight:195.602 g/mol
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together