3-O-Benzyl-4-(hydroxymethyl-1,2-O-isopropylidene)-α-D-erythropentofuranose 97%
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: 3-O-Benzyl-4-(hydroxymethyl-1,2-O-isopropylidene)-alpha-D-erythropentofuranose, [(3aR,6S,6aR)-6-(benzyloxy)-5-(hydroxymethyl)-2,2-dimethyl-tetrahydro-2H-furo[2,3-d][1,3]dioxol-5-yl]methanol, 3-O-Benzyl-4-(hydroxymethyl)-1,2-O-isopropylidene-ALPHA-D-ribofuranose, ((3aR,6S,6aR)-6-(Benzyloxy)-2,2-dimethyltetrahydrofuro[2,3-d][1,3]dioxole-5,5-diyl)dimethanol
CAS Number:63593-03-3
:
Formula:C₁₆H₂₂O₆
MDL Number:MFCD08704144
Molecular Weight:310.342 g/mol
:GHS07
Storage Temperature:Refrigerator
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together