4-Methoxy-2,2-dimethyl-4-oxobutanoic acid 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:54043-71-9
:
Formula:C₇H₁₂O₄
MDL Number:MFCD16619681
Molecular Weight:160.168 g/mol
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together