7-Bromo-1-hydroxynaphthalene 95%
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: 7-Bromonaphthalen-1-ol
CAS Number:91270-69-8
:
Formula:C₁₀H₇BrO
MDL Number:MFCD17012439
Molecular Weight:223.066 g/mol
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together