Methyl 3-bromo-4-sulphamoylbenzoate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:89978-60-9
:
Formula:C₈H₈BrNO₄S
MDL Number:MFCD21602570
Molecular Weight:294.122 g/mol
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together