[3-(Methacryloylamino)propyl]dimethyl(3-sulphopropyl)ammonium hydroxide inner salt 97%
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: 3-(Dimethyl{3-[(2-methylacryloyl)amino]propyl}ammonio)propane-1-sulphonate, 3-sulphopropyldimethyl-3-methacrylamidopropylammoniuminnersalt
CAS Number:5205-95-8
:
Formula:C₁₂H₂₄N₂O₄S
MDL Number:MFCD00080642
Molecular Weight:292.395 g/mol
Melting Pt:190 °C (dec.)
Storage Temperature:Ambient
EINECS:226-003-9
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together