Order Entry
Slovenia
ContactUsLinkComponent
4-Bromo-6-methyl-1H-pyrrolo[2,3-b]pyridine ≥98%
4-Bromo-6-methyl-1H-pyrrolo[2,3-b]pyridine ≥98%
Catalog # APOSOR510156-100MG
Supplier:  Apollo Scientific
CAS Number:  
4-Bromo-6-methyl-1H-pyrrolo[2,3-b]pyridine ≥98%
Catalog # APOSOR510156-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440
  • Environmentally Preferable:

Specifications

About this item

 :
 :211.059 g/mol
Storage Temperature:Ambient
CAS Number:1000340-58-8
Formula:C₈H₇BrN₂
MDL Number:MFCD09880145