Order Entry
Slovenia
ContactUsLinkComponent
Boc-2-Cyclopropylglycine
Boc-2-Cyclopropylglycine
Catalog # APOSOR451165-5G
Supplier:  Apollo Scientific
CAS Number:  
Boc-2-Cyclopropylglycine
Catalog # APOSOR451165-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173
  • Environmentally Preferable:

Specifications

About this item

Synonyms: 2-((tert-Butoxycarbonyl)amino)-2-cyclopropylacetic acid, Boc-DL-Cyclopropyl-Gly-OH
 :
 :215.246 g/mol
Storage Temperature:Ambient
CAS Number:54256-41-6
Formula:C₁₀H₁₇NO₄
MDL Number:MFCD01861823