Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Chloro-pyrazine-2-carboxylic acid ethyl ester
3-Chloro-pyrazine-2-carboxylic acid ethyl ester
3-Chloro-pyrazine-2-carboxylic acid ethyl ester
3-Chloro-pyrazine-2-carboxylic acid ethyl ester
APOSOR346138-5G
:
Apollo Scientific
:
3-Chloro-pyrazine-2-carboxylic acid ethyl ester
APOSOR346138-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
Environmentally Preferable:
:
Ethyl 3-chloropyrazine-2-carboxylate
:
:
186.596 g/mol
:
Ambient
:
655247-45-3
:
C₇H₇ClN₂O₂
:
MFCD07371661