3-Bromo-2-hydrazinopyridine
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: 3-Bromo-2-hydrazinylpyridine
CAS Number:54231-41-3
:
Formula:C₅H₆BrN₃
MDL Number:MFCD00490113
Molecular Weight:188.025 g/mol
:GHS07
Storage Temperature:Refrigerator
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together