Graduated wide neck square bottles, PE, with screw cap and undercap
Supplier: KARTELL
Certificates
About this item
These high quality graduated, wide neck square bottles are made of PE (main body and under cap) and supplied with a PP cap.
- Suitable for foodstuff contact according to EC Regulation 10/2011
- Translucent
- Tagging points on shoulder and cap ensures tamper-evidence by means of tags and seals
Graduated space saving bottles, ideal for long-term storage of solid or liquid samples. Sloping shoulders reduce tendency for residue deposits. Moulded-in graduations.
Certifications: Meets EC Regulation 10/2011. Migration Test Compliance Declaration available.
Frequently Bought Together