Potassium trifluoro(trifluoromethyl)borate
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Potassium (trifluoromethyl)trifluoroborate
CAS Number:42298-15-7
:
Formula:CHBF₆
MDL Number:MFCD09038544
Molecular Weight:175.91 g/mol
Melting Pt:>300 °C
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to yellow to brown Crystalline powder or solid
- Infrared spectrum:Conforms
- NMR (19F-NMR):≥96 %
Specification Test Results
Frequently Bought Together