Dextrose solution 50% (w/w), Biotech Reagent, J.T.Baker®
Catalog # 0890-09
Supplier: Avantor
CAS Number:
Specifications
- Pk:200 L
- Seq:5549
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay (w/w):49 - 51 %
- Osmolality, mOsm/kg:317 - 393
- Endotoxin Concentration (EU/g):≤1
Specification Test Results
About this item
Synonyms: Corn sugar, ᴅ-(+)-Glucose, Glucose, D-(+)-Dextrose, (2R,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal, Glucolin, Dextrose, D-Glucose
:
:180.16 g/mol
CAS Number:50-99-7
:492-62-6
Formula:C₆H₁₂O₆
MDL Number:MFCD00148912
Melting Pt:146
Merck Index:14,04459
:Management of Change (MOC) category = R