Reagent reservoirs, disposable
Supplier: BIOTIX
Certificates
About this item
These disposable reagent reservoirs are designed for use with multi-channel pipetting applications and can accommodate up to 12-channel pipettes. Users can choose from opaque white polyvinyl chloride (PVC) or high clarity polystyrene (PS) in several packaging configurations.
- Sterile or non sterile
- Angled sides and a V-shaped bottom allow maximum recovery of reagents
- All reservoirs feature internal graduated volume lines and pour-spouts on the corners
- Certified free of DNA, DNase, RNase and endotoxins (pyrogens)
Manufactured under stringent controls in an ISO 9001 certified facility. Every lot is tested assuring maximum quality and compatibility. All raw materials are traceable.
Manufactured under stringent controls in an ISO 9001 certified facility. Every lot is tested for precision assuring maximum quality and compatibility. All raw materials are traceable.
Packaging: Reagent reservoirs are available pre-sterile, non sterile, bulk and individually wrapped.
Frequently Bought Together