Order Entry
Slovenia
ContactUsLinkComponent
1-Propanesulphonic acid 95%
1-Propanesulphonic acid 95%
Certificates
undefined
1-Propanesulphonic acid 95%

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    Clear brown to dark brown Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4380 to 1.4410 (20°C, 589 nm)
  • Titration with NaOH:
    ≥94 %

Specification Test Results

Related Information

Frequently Bought Together