Order Entry
Slovenia
ContactUsLinkComponent
Tetraethylammonium fluoride hydrate 98%
undefined
Tetraethylammonium fluoride hydrate 98%
  • Environmentally Preferable:
  • Appearance:
    White to beige Crystals or adhering crystals or crystalline solid
  • Infrared spectrum:
    Conforms
  • Water:
    ≤28 % (K.F.)
  • Titration with HClO4:
    ≥97.5 % (On dry substance)
Frequently Bought Together