Bucket Liners
The unique LDPE/nylon laminate is a good barrier material for transporting raw materials or to line buckets containing disinfectants or other cleaning solutions.
- Laminate is 75% LDPE, 25% nylon
- Excellent for disinfectants and cleaning solutions
- Ideal for product parts transport
Designed for cleanrooms and controlled environments. Available as sterile and non-irradiated.
: Individually double packed, packed in a case of 24/50/200.
- Catalog No:
- MICRBKL99-3521
- MICRBKL7-3119
- MICRBKL99-3521IR
- MICRBKL-2430IR
- MICRBKL-2424IR
- MICRBKL1-1818IR
- Environmentally Preferable:
- Waste Reducing
- Waste Reducing
Frequently Bought Together