Supelco® PESTANAL® Organic Reference Standard, Dodin
45466-250MG
: Merck
:
- Pk:250 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:2439-10-3
- Description:Dodin
- Environmentally Preferable:
- Storage conditions:2...30 °C
- Hazard class description:H302 - H315 - H319 - H410
- Shipping conditions:STD
- MDL No.:MFCD00034998
- M-Clarity quality level:MQ100
- Synonym(s):1-Dodecyl-guanidiniumacetat
- Formula:C₁₅H₃₃N₃O₂
- Molecular weight:287,44
- GHS signal: GHS07, GHS09
: P264 - P270 - P273 - P301 + P312 - P302 + P352 - P305 + P351 + P338