Order Entry
Slovenia
ContactUsLinkComponent
Genomic DNA purification kit, Fermentas
Genomic DNA purification kit, Fermentas
Catalog # K0512
CAS Number:  
undefined
Genomic DNA purification kit, Fermentas
Catalog # K0512
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Application:
  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Total DNA
  • Sample type:
    Plant samples
    Bacteria
    Blood
    Biofluids (serum, plasma, saliva)
    Yeast/Fungi
    Cultured cells
    Human/animal cells and tissues
  • Target molecule:
    DNA
  • Description:
    Genomic DNA purification kit, 100 preps
  • Environmentally Preferable:
  • Description:
    Genomic DNA purification kit

Specifications

About this item

DNA purification kit is a simple and rapid system for high quality genomic DNA purification from various sources including: whole blood, serum, cell lines, bacterial cells, plant, and mammalian tissues.

  • Efficient - 2 to 10 µg of genomic DNA from 0,2 ml of blood
  • Fast - procedure takes approximately 20 minutes
  • Universal - purifies genomic DNA from various sources
  • Flexible - easy to scale up and down
  • Safe - does not involve phenol extraction

High molecular weight genomic DNA purified with the kit is suitable for direct use in all common molecular biology applications: PCR, FastDigest and conventional restriction digestion, cloning, DNA sequencing, and southern blot analysis.

The kit is based on selective detergent-mediated DNA precipitation from crude lysate. The entire procedure is rapid only 20 to 25 minutes with a typical yield of 2 to 10 µg genomic DNA from 0,2 ml of blood.

Applications include purification of high-quality genomic DNA from wWhole blood or serum, cell culture, plant tissues, mammalian tissues, epithelium samples, bacteria includes lysis solution precipitation solution NaCl solution detailed protocol.

Delivery: The kit contains lysis solution (40 ml), precipitation solution (10X) (8 ml), Sodium chloride (1,2 M) (10 ml). Store the kit at room temperature.
Caution: For research use only. Not for use in diagnostic procedures.