Order Entry
Slovenia
ContactUsLinkComponent
 
Aluminium foil, roll
Catalog # KORF70237
Supplier:  Korff
undefined
Aluminium foil, roll
Catalog # KORF70237
Supplier:  Korff

Specifications

  • Width:
    150 mm
  • Thickness:
    0,020 mm
  • Environmentally Preferable:
  • Pk:
    1 Roll

Specifications

About this item

This aluminum foil is packed on rolls of about 5 kg.