Order Entry
Slovenia
ContactUsLinkComponent
Gel documentation system, G:BOX Mini
Gel documentation system, G:BOX Mini
Certificates
undefined
Gel documentation system, G:BOX Mini
Specifications
  • Catalog No:
  • SYNOGBOX-MINI-6-E
  • SYNOGBOX-MINI-9-E
  • Model:
  • G:BOX-mini-6
  • G:BOX-mini-9
  • Camera:
  • 16 Bit, 65536 Grey Scale
  • 16 Bit, 65536 Grey Scale
  • Resolution:
  • 6 megapixels
  • 9 megapixels
  • Lighting:
  • Epi LED white
  • Epi LED white
  • Image area:
  • 150×120 mm (Max.)
    100×80 mm (Min.)
  • 150×120 mm (Max.)
    100×80 mm (Min.)
  • Lens:
  • F0,95 (motor driven)
  • F0,95 (motor driven)
  • W×D×H:
  • 400×520×640 mm
  • 400×520×640 mm
  • Weight:
  • 40 kg
  • 40 kg