Empty FPLC columns, G-TRAP™
Supplier: G-Biosciences
Certificates
About this item
These empty columns are designed for fast protein liquid chromatography.
- Frit material is ultra-high molecular weight polyethylene
Columns are made of polypropylene and come in 1 ml and 5 ml sizes.
Delivery: Includes column tube, sealing plug, sealing sleeve, top stop plug and bottom stop plug, top (large) frit, bottom (small) frit (inserted in column).
Specifications
- Catalog No:
- GENO786-1291
- GENO786-1290
- Max. pressure:
- 3 bar
- 3 bar