Order Entry
Slovenia
Orders LinkContactUsLinkComponent
Magnetic separation rack, MagJET

Magnetic separation rack, MagJET

Magnetic separation rack, 1,5 ml, 12
Catalog # PIERMR02
Magnetic separation rack, MagJET
Catalog # PIERMR02
Magnetic separation rack, MagJET

Magnetic separation rack, 1,5 ml, 12

Price
482,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents

This separation rack is a magnetic separation device intended for low-throughput magnetic bead manipulation procedures such as magnetic bead-based nucleic acid isolation.

  • Contains a powerful permanent neodymium magnet for instantaneous bead collection from the solution
  • Specially designed for robust performance with different sample volumes
  • Stable and tight magnetic bead capture on microcentrifuge tube wall
  • Highly efficient nucleic acid purification in combination with Thermo Scientific MagJET magnetic-bead based kits

The MagJET separation rack contains a high-performance permanent neodymium magnet embedded in a plastic separation unit frame. In combination with MagJET magnetic beads the separation aack allows fast and efficient magnetic bead-based separation procedures.

Applications: Affinity purification using magnetic particles.

Specifications
  • For tubes:
    1,5 ml
  • No. of Holes:
    12
  • Description:
    Magnetic separation rack
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.