Latex gloves, SemperCare®
Supplier: SEMPERIT TECHN PROD
Certificates
About this item
These Sempercare® disposable latex gloves are ideal for safely handling instruments and for sensitive operations.
- Powdered for easy donning
- High wearing comfort
- Micro rough surface for good grip, even in moist environments
Primarily intended for medical use, to protect both patient and user against cross-contamination.
Certifications: ASTM D3578; EN 455 1-4, EN 420, EN 374-1, -2, EN 374-3 / EN 16523-1; AQL 1,5
Food contact approved.
Food contact approved.
Frequently Bought Together