JP-8 military fuel standard
REST31254
: Restek
:
- Concentration:50000 μg/ml in methylene chloride
- Pk:1 mL
- CAS No.:94114-58-6
- Description:JP-8 military fuel standard
- Environmentally Preferable:
- Pk Info:1 ml/ampoule
: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.