Order Entry
Slovenia
ContactUsLinkComponent
DNA marker, pBR322 DNA/BsuRI (HaeIII) marker
DNA marker, pBR322 DNA/BsuRI (HaeIII) marker
Catalog # SM0271
CAS Number:  
undefined
DNA marker, pBR322 DNA/BsuRI (HaeIII) marker
Catalog # SM0271
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    pBR322 DNA/BsuRI (HaeIII) marker, 50 µg
  • Environmentally Preferable:
Specifications
About this item
pBR322 DNA/BsuRI (HaeIII) marker is suitable for sizing and approximate quantification of small linear double-stranded DNA fragments in agarose and non denaturing polyacrylamide gels.
  • Agarose gel compatible
  • Sharp bands
Delivery: Supplied with loading dye for sample DNA.
Caution: For research use only. Not for use in diagnostic procedures.