Sodium L(+)-ascorbate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Sodium ascorbate, L(+)-Ascorbic acid sodium salt, L-Ascorbic acid sodium salt, Sodium (R)-2-((S)-1,2-dihydroxyethyl)-4-hydroxy-5-oxo-2,5-dihydrofuran-3-olate, Acidum ascorbicum sodium salt
CAS Number:134-03-2
:
Formula:C₆H₇NaO₆
MDL Number:MFCD00082340
Density:1.799 g/cm³ (20 °C)
Molecular Weight:198.11 g/mol
Melting Pt:219
:M-Clarity™ quality level = MQ300
Storage Temperature:Ambient
EINECS:205-126-1
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together