Order Entry
Slovenia
ContactUsLinkComponent
 
Ammonium chloride 99+%, pure
 
undefined
Ammonium chloride 99+%, pure
 

 

  • Environmentally Preferable:

 

 

  • Appearance:
    White Crystalline powder and/or chunks
  • Titration Argentometric:
    ≥99.0 %
  • Water:
    ≤0.1 % (K.F.)
  • Sulfated ash:
    ≤0.3 %
  • Iron (Fe):
    ≤10 ppm
  • Insoluble matter:
    ≤0.02 %

 

 

Frequently Bought Together