Aluminium chloride, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Aluminum chloride
CAS Number:7446-70-0
:
Formula:AlCl₃
MDL Number:MFCD00003422
Boiling Pt:180
Molecular Weight:133.34 g/mol
Melting Pt:180…181 °C
ADR:8 II
Merck Index:13,00335
:M-Clarity™ quality level = MQ200
EINECS:231-208-1
UN:1726
:GHS05
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together