Potassium manganate
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10294-64-1
:
Formula:K₂MnO₄
MDL Number:MFCD00043089
Molecular Weight:197.13 g/mol
Melting Pt:190 °C (dec.)
ADR:5.1 III
Merck Index:12,07810
EINECS:233-665-2
UN:1479
:GHS03
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together