1,4-Cyclohexadiene 97% stabilised
: Cyclohexa-1,4-diene
:628-41-1
:
:C₆H₈
:MFCD00001535
:81…82 °C (760 mmHg)
:–49.2 °C
:0.84 g/cm³
:80.13 g/mol
:–6 °C
:Refrigerator
:211-043-1
:3295
:3 II
:GHS02
- Environmentally Preferable:
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4715 to 1.4735 (20°C, 589 nm)
- GC:≥96.0 %
- Stabilizer:0.1 % BHT
Frequently Bought Together