Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,3,3',5',6-Pentachlorobiphenyl (PCB No. 113)
2,3,3',5',6-Pentachlorobiphenyl (PCB No. 113)
2,3,3',5',6-Pentachlorobiphenyl (PCB No. 113)
2,3,3',5',6-Pentachlorobiphenyl (PCB No. 113)
Catalog # PROOCIL-PCB-113-C
Supplier:
LGC Standards PROMOCHEM
CAS Number:
Expand image
2,3,3',5',6-Pentachlorobiphenyl (PCB No. 113)
Catalog # PROOCIL-PCB-113-C
Supplier:
LGC Standards PROMOCHEM
CAS Number:
About this item
Product Details
Molecular Weight:
326.44 g/mol
Product Details