2,2-Diphenylpropane, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Propane-2,2-diyldibenzene
CAS Number:778-22-3
:
Formula:(C₆H₅)₂C(CH₃)₂
MDL Number:MFCD00008794
Boiling Pt:281
Melting Pt:29
Density:0.998 g/cm³ (20 °C)
Molecular Weight:196.29 g/mol
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:212-292-9
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together