Order Entry
Slovenia
ContactUsLinkComponent
Benzyl alcohol ≥97%, TECHNICAL
Benzyl alcohol ≥97%, TECHNICAL
Catalog # 20804.366
Supplier:  VWR Chemicals
CAS Number:  
undefined
Benzyl alcohol ≥97%, TECHNICAL
Catalog # 20804.366
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    5 L
  • Seq:
    5190
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Assay:
    Min. 97 %

Specification Test Results

About this item

Synonyms: Benzenemethanol, Phenylmethanol, Phenylcarbinol
 :
 :108.14 g/mol
Boiling Pt:205
EINECS:202-859-9
 :GHS07
Melting Pt:–15,3 °C
Storage Temperature:Ambient
CAS Number:100-51-6
Formula:C₆H₅CH₂OH
MDL Number:MFCD00004599
Density:1.05 g/cm³ (20 °C)
Flash Pt:101
Merck Index:13,01126