Order Entry
Slovenia
Orders LinkContactUsLinkComponent
ω-Bromo-3-methoxyacetophenone, Sigma-Aldrich®

ω-Bromo-3-methoxyacetophenone, Sigma-Aldrich®

Supplier:   Merck
ω-Bromo-3-methoxyacetophenone, Sigma-Aldrich®
Supplier:  Merck
Certificates
ω-Bromo-3-methoxyacetophenone, Sigma-Aldrich®

2 Options Available Below

Product Details & Documents
Synonyms:  2-Bromo-3'-methoxyacetophenone, Bromomethyl m-methoxyphenyl ketone, 3-Methoxyphenacyl bromide, 2-Bromo-1-(3-methoxyphenyl)ethanone, 2-Bromo-1-(3-methoxyphenyl)ethan-1-one, α-Bromo-3-methoxyacetophenone
  • CAS Number:5000-65-7
  • Formula:C₉H₉BrO₂
  • MDL Number:MFCD00000199
  • Molecular Weight:229.07 g/mol
  • Boiling Pt:285 °C (1013 hPa)
  • Melting Pt:63…64.3 °C
  • Storage Temperature:Refrigerator
  • EINECS:225-666-1
  • UN:3261
  • ADR:8,II
  •  :M-Clarity™ quality level = MQ100
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g - 94,40 €
    94,40 €
    1 * 10 g
       94,40 €

  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 208,00 €
    208,00 €
    1 * 25 g
       208,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.