Benzal bromide, Sigma-Aldrich®
SIAL123560-100G
: Merck
:
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
- Environmentally Preferable:
: α,α-Dibromotoluene, (Dibromomethyl)benzene
:
:249.93 g/mol
:210-543-7
:2927
:GHS06
:GHS05
:Ambient
:618-31-5
:(C₆H₅)N(CH₂Br)₂
:156
:0…1 °C
:1.51 g/cm³ (15 °C)
:6.1 II
:M-Clarity™ quality level = MQ100