Resveratrol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: trans-3,4,5-Trihydroxystilbene, (E)-5-(4-Hydroxystyryl)benzene-1,3-diol
CAS Number:501-36-0
:
Formula:C₁₄H₁₂O₃
MDL Number:MFCD00133799
Density:1.36 g/cm³ (20 °C)
Molecular Weight:228.25 g/mol
Melting Pt:260
Merck Index:12,08326
:M-Clarity™ quality level = MQ200
Storage Temperature:Freezer
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together