2,6-Dimethoxyphenylboronic acid
Supplier: Thermo Scientific
Certificates
About this item
CAS Number:23112-96-1
:
Formula:C₈H₁₁BO₄
MDL Number:MFCD01318987
Molecular Weight:181.98 g/mol
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together