Dithiothreitol (DTT, Cleland's reagent) ≥98%
Supplier: VWR Chemicals
Certificates
About this item
Synonyms: cis-2,3-Dihydroxy-1,4-butanedithiol, (R*,R*)-1,4-Dimercapto-2,3-butandiol, DTT, DL-1,4-Dithiothreitol
CAS Number:3483-12-3
:27565-41-9
Formula:C₄H₁₀O₂S₂
MDL Number:MFCD00004877
Boiling Pt:< 125 °C (1013 hPa)
Molecular Weight:154.25 g/mol
Melting Pt:42…44 °C
Flash Pt:>110 °C (open cup)
EINECS:222-468-7
:GHS07
Storage Temperature:Refrigerator
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay:Min. 98 %
- IR Spectrum:Passes test
- Melting point:40 - 43 °C
- Heavy metals (as Pb):Max. 10 ppm
- Fe (Iron):Max. 10 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Number250 ml Plastic bottle for solids DIN 60 (ZPACK13250250)
Packaging Information
441496P_1,4-DITHIOTHREITOL_BSE_REV0.pdf
BSE/TSE Certificates
spec_VWRC441494N spec_VWRC441496P spec_VWRC441496P
Product specification
Frequently Bought Together