1,6-Diaminohexane ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 1,6-Hexanediamine, Hexamethylenediamine
CAS Number:124-09-4
:
Formula:C₆H₁₆N₂
MDL Number:MFCD00008243
Boiling Pt:199…204 °C
Melting Pt:39…43 °C
Density:0.840
Molecular Weight:116.21 g/mol
Flash Pt:85 °C (185 °F)
UN:2280
ADR:8 III
Merck Index:13,04713
EINECS:204-679-6
:GHS05
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together