Nickel(II) nitrate hexahydrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:13478-00-7
:
Formula:Ni(NO₃)₂·6H₂O
MDL Number:MFCD00149805
Boiling Pt:137
Melting Pt:56.7
Density:2.05 g/cm³ (20 °C)
Molecular Weight:290.79 g/mol
EINECS:236-068-5
UN:2725
ADR:5.1 III
Merck Index:12,06596
:M-Clarity™ quality level = MQ200
:GHS03
:GHS05
:GHS08
:GHS07
:GHS09
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together