di-Ammonium oxalate monohydrate 99.5-101.0%, AnalaR NORMAPUR® analytical reagent
: Ammonium oxalate monohydrate, Ammonium oxalate-1-hydrate, Oxalic acid diammonium salt monohydrate, Oxalic acid ammonium salt monohydrate, Ammonium ethanedioate monohydrate, Ammonium Oxalate, Monohydrate
:6009-70-7
:
:(NH₄)₂C₂O₄·H₂O
:MFCD00149694
:70
:∼1.5 g/cm³ (20 °C)
:142.11 g/mol
:Ambient
:214-202-3
:3288
:6.1 III
:13,00541
:GHS07
- Environmentally Preferable:
- Assay:99.5 - 101.0 %
- Heavy metals (as Pb):Max. 5 ppm
- Ignition residue (SO4):Max. 0.02 %
- Insolubility in water:Max. 50 ppm
- Cl (Chloride):Max. 20 ppm
- NO3 (Nitrate):Max. 20 ppm
- SO4 (Sulphate):Max. 20 ppm
- Ca (Calcium):Max. 20 ppm
- Fe (Iron):Max. 5 ppm
- K (Potassium):Max. 100 ppm
- Mg (Magnesium):Max. 10 ppm
- Na (Sodium):Max. 50 ppm
Frequently Bought Together