Polyethylene glycol 8,000
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: PEG 8,000, Ethylene glycol polymer 8000, PEG 8000
CAS Number:25322-68-3
:MFCD00081839
Formula:H(OCH₂CH₂)nOH
MDL Number:MFCD01779615
Boiling Pt:250
Melting Pt:55…63 °C
Density:1.125 g/cm³ (20 °C)
Molecular Weight:8000 g/mol
Merck Index:14,7568
Storage Temperature:Ambient
EINECS:500-038-2
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together