1,2-Cyclohexanediol cis- and trans-mixture ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Cyclohexane-1,2-diol
CAS Number:931-17-9
:
Formula:C₆H₁₂O₂
MDL Number:MFCD00003861
Boiling Pt:118…120 °C (10 mmHg)
Melting Pt:72…76 °C
Density:0.948 g/cm³ (20 °C)
Molecular Weight:116.16 g/mol
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
EINECS:213-229-8
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together