Order Entry
Slovenia
Orders LinkContactUsLinkComponent
Automated stainer, Cellstain® 23

Automated stainer, Cellstain® 23

Automatic stainer incl. 23 staining troughs with lid and slide holder, Cellstain® 23, 30
Catalog # THARJC800-30
Supplier:   THARMAC GmbH
Automated stainer, Cellstain® 23
Catalog # THARJC800-30
Supplier:  THARMAC GmbH
Automated stainer, Cellstain® 23
Automated stainer, Cellstain® 23

Automatic stainer incl. 23 staining troughs with lid and slide holder, Cellstain® 23, 30

Price
14 200,00 €
Log in for availability
1 * 1 items
Product Details & Documents

The CellStain 23 features programming unit for individually adjustable steps in staining, rinsing and drying stations and implements of different stainings due to forwards and backwards movement of slide holder. It contains storage place for 20 staining programs up to 30 steps each with password protected access.

  • Low transfer of reagents through the slide holder
  • Small format fits under any existing fume hood
  • Optional drying station with programmable temperature

Delivery information: Supplied with 23 staining troughs (500 ml) with lid, one slide holder for up to 30 slides and one shortened trough for loading and unloading.

Specifications
  • Model:
    Cellstain® 23
  • No. of slides:
    30
  • Description:
    Automatic stainer incl. 23 staining troughs with lid and slide holder
  • Cat. No.:
    THARJC800-30
  • No. of programs:
    20
  • W×D×H:
    1600×280×350 mm
  • Weight:
    25 kg
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Adjustable slide dryer (40...80 °C)
    Adjustable slide dryer (40...80 °C)
    THARJC653
    Log in for availability
    1 * 1 items - 712,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Charcoal filter for fume hoods
    Charcoal filter for fume hoods
    THARJC690AK
    Log in for availability
    1 * 1 items - 89,70 €

Product Information

  • Catalog #
    Availability
    Price
  • Staining trough with lid, 500 ml
    Staining trough with lid, 500 ml
    THARJC621-1
    Log in for availability
    1 * 1 items - 91,70 €

Product Information

  • Catalog #
    Availability
    Price
  • Heating module with one heating trough
    Heating module with one heating trough
    THARJC702-1
    Log in for availability
    1 * 1 items - 2 570,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Slide holder (for up to 30 slides)
    Slide holder (for up to 30 slides)
    THARJC612-1
    Log in for availability
    1 * 1 items - 234,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Fume hood with activated charcoal filter
    Fume hood with activated charcoal filter
    THARJC690-23
    Log in for availability
    1 * 1 items - 2 770,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Water rinse station
    Water rinse station
    THARJC654
    Log in for availability
    1 * 1 items - 379,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.