Aluminium oxide 20% (w/w) in 2-propanol, nanoparticle dispersion <50 nm (DLS), Sigma-Aldrich®
: Merck
: Aluminium(III) oxide, Alumina C, Aluminum oxide
:1344-28-1
:11092-32-3
:Al₂O₃
:MFCD00003424
:2980
:2045
:0.79 g/cm³ (25 °C)
:101.96 g/mol
:12 °C
:1219
:3 II
:13,00355
:M-Clarity™ quality level = MQ100
:GHS02
:GHS07
- Environmentally Preferable:
- Loading, Dispersion :19.0 - 21.0% wt
- Dispersion pH:7.5 - 9.5
- Particle Size (ICP):≤50 nm
- ICP Major Analysis (Aluminium Component):Confirmed
- Purity (≥99.9% Based on Trace Metals Analysis):Meets Requirements
- Trace Metal Analysis:≤1000.0 ppm
Frequently Bought Together