Order Entry
Slovenia
ContactUsLinkComponent
Anti-TRAIL-R1 Mouse Monoclonal Antibody [clone: DJR1]
Anti-TRAIL-R1 Mouse Monoclonal Antibody [clone: DJR1]
Catalog # ENZOALX8049120100
Supplier:  ENZO LIFE SCIENCES
New Product
Anti-TRAIL-R1 Mouse Monoclonal Antibody [clone: DJR1]
Catalog # ENZOALX8049120100
Supplier:  ENZO LIFE SCIENCES
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    TNF receptor superfamily member 10a
  • Antigen symbol:
    TRAIL-R1
  • Clonality:
    Monoclonal
  • Clone:
    DJR1
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
  • Western blot:
    Yes
  • Seq:
    02
  • Environmentally Preferable:
  • Form:
    Liquid
  • Antigen synonyms:
    TRAILR1|DR4|APO2|CD261|TRAILR-1|MGC9365
  • Storage buffer:
    PBS, pH 7,2. containing 0,09% sodium azide.
  • Storage temperature:
    4 °C
  • Shipping temperature:
    Blue Ice Not Frozen
  • Immunogen:
    Recombinant human TRAIL-R1 extracellular domain IgG1 Fc fusion protein.
  • Purification:
    Purified by affinity chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

DR4 is 56kDa member 10A of the TNFR superfamily (TNFRSF10A), also known as TRAIL-R1, Apo-2, and CD261. It is expressed at low levels by activated T cells and some tumors. After TRAIL engagement, DR4 (TRAIL-R1), through activation of NF-κB, induces apoptosis in the TRAIL ligated cell.

Type: Primary
Antigen: TRAIL-R1
Clonality: Monoclonal
Clone: DJR1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human