Silt density index kit
Catalog # LOVI56K001901
Supplier: Lovibond
CAS Number:
Specifications
- Pk:1
- Description:Silt density index (SDI) test kit
- Environmentally Preferable:
Specifications
About this item
The Silt Density Index (SDI) is a measure of the solids or particulate content of water and is of specific importance when commissioning reverse osmosis plant.
- SDI calculations in just 15 minutes
- Ideally suited for reverse osmosis plants
- Enables calculation of membrane fouling time, backwash frequency and membrane lifespan
The kit utilises a membrane/flow meter system to enable SDI calculations in just 15 minutes. Sufficient membranes included for 100 tests.